missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 2 Antibody (TGFB2/1679) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-27016JF669
This item is not returnable.
View return policy
Description
TGF-beta 2 Monoclonal antibody specifically detects TGF-beta 2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TGF-beta 2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2 | |
| Recombinant full-length human TGF-beta 2 protein | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Tumor Suppressors | |
| 7042 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| TGFB2/1679 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction