missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 3 Antibody (Brigham and Womens anti-LAP), DyLight 350, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28029UV
This item is not returnable.
View return policy
Description
TGF-beta 3 Monoclonal antibody specifically detects TGF-beta 3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| TGF-beta 3 | |
| Monoclonal | |
| DyLight 350 | |
| ARVD, FLJ16571, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3 | |
| LAP | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Tumor Suppressors | |
| 7043 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| Brigham and Womens anti-LAP | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion