missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TGF-beta 3 Antibody (TGFB3/4801), Novus Biologicals™
Mouse Monoclonal Antibody
€ 245.00 - € 458.85
Specifications
| Antigen | TGF-beta 3 |
|---|---|
| Clone | TGFB3/4801 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043998
|
Novus Biologicals™
NBP3-26735-100ug |
100 μg |
€ 486.00 € 458.85 / 100µg Save € 27.15 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043518
|
Novus Biologicals™
NBP3-26735-20ug |
20 μg |
€ 245.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGF-beta 3 Monoclonal antibody specifically detects TGF-beta 3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TGF-beta 3 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 7043 | |
| IgG2 κ | |
| Protein A or G purified |
| TGFB3/4801 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ARVD, FLJ16571, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3 | |
| Recombinant fragment (around aa50-250) of human TGF-beta 3 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title