Learn More
PeproTech TGF beta-3 Polyclonal Antibody, PeproTech®, Invitrogen™

Description
AA Sequence of recombinant protein: ALDTNYCFRN LEENCCVRPL YIDFRQDLGW KWVHEPKGYY ANFCSGPCPY LRSADTTHST VLGLYNTLNP EASASPCCVP QDLEPLTILY YVGRTPKVEQ LSNMVVKSCK CS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TGF-beta3. Anti-Human TGF-beta3-specific antibody was purified by affinity chromatography employing an immobilized Human TGF-beta3 matrix. Western Blot: To detect Human TGF-beta3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human TGF-beta3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Specifications
Specifications
| Antigen | TGF beta-3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | TGFB3 |
| Gene Accession No. | P10600 |
| Gene Alias | ARVD; ARVD1; FLJ16571; H-TGF-b-3; LAP; Latency-associated peptide; LDS5; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; Tgfb3; TGF-B3; Tgfb-3; TGF-beta3; TGF-beta-3; Transforming growth factor; transforming growth factor beta 3; transforming growth factor beta precursor; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3; transforming growth factor-beta3 |
| Gene Symbols | TGFB3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.