missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta Antibody (TGFB/510), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08739G
This item is not returnable.
View return policy
Description
TGF-beta Monoclonal antibody specifically detects TGF-beta in Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey samples. It is validated for ELISA, Functional Assay
Specifications
| TGF-beta | |
| Monoclonal | |
| DyLight 488 | |
| CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| Full length recombinant human TGF-beta protein | |
| 100 μg | |
| Cytokine Research, Extracellular Matrix | |
| 7040 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Functional Assay | |
| TGFB/510 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction