missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta Antibody (TGFB/7230), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24125AF532
This item is not returnable.
View return policy
Description
TGF-beta Monoclonal antibody specifically detects TGF-beta in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TGF-beta | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| Recombinant fragment (around aa200-390) of human TGF-beta protein | |
| 0.1 mL | |
| Cytokine Research, Extracellular Matrix | |
| 7040 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| TGFB/7230 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction