missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta RIII Antibody (MM0057-5G9), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11920H
This item is not returnable.
View return policy
Description
TGF-beta RIII Monoclonal specifically detects TGF-beta RIII in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TGF-beta RIII | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TGFBR3 | |
| Human recombinant TGF-beta RIII (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| In direct ELISAs, this antibody does not cross-react with recombinant human TGF-beta RI, and TGF-beta RII. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0057-5G9 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| betaglycan, betaglycan proteoglycan, BGCAN, TGF-beta receptor type 3, TGF-beta receptor type III, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa) | |
| Mouse | |
| Protein G purified | |
| Angiogenesis | |
| 7049 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction