missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGFA Mouse anti-Human, Clone: TG86, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14613.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against synthetic peptide of human TGFA.
- High specificity
- High reactivity
Specifications
| TGFA | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| P01135 | |
| TGFA | |
| A synthetic peptide corresponding to amino acids 34-43 of human TGFA. | |
| 100 μg | |
| Primary | |
| This antibody reacts with the TGFA and shows no cross-reaction with EGF and the neuropeptide synenkephalin. | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| TG86 | |
| Mouse monoclonal antibody raised against synthetic peptide of human TGFA. | |
| In 10 mM PBS. | |
| TFGA | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 7039 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction