missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGFBR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120051
This item is not returnable.
View return policy
Description
Positive test controls include: 293T, Mouse testis, Rat testis, Rat brain. The target is usually found in the following locations: Cell junction, Cell membrane, Single-pass type I membrane protein, tight junction. Immunogen sequence: EEDPSLDRPF ISEGTTLKD.
TGFBR1 is a transmembrane serine/threonine kinase in the TGFBR1 superfamily, which also includes ACVRs and BMPRs. Upon binding TGF-b, TGFBR2 dimerizes with and activates TGFBR1 via phosphorylation leading to downstream phosphorylation of SMADs by TGFBR1. TGFBR1 forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF-beta signal from the cell surface to the cytoplasm. The encoded protein is a serine/threonine protein kinase. Mutations in this gene have been associated with Loeys-Dietz aortic aneurysm syndrome (LDAS).
Specifications
| TGFBR1 | |
| Polyclonal | |
| Unconjugated | |
| TGFBR1 | |
| AAT5; activin A receptor type II-like kinase, 53kDa; activin A receptor type II-like protein kinase of 53kD; Activin receptor-like kinase 5; ACVRLK4; ALK5; Alk-5; AU017191; ESK2; ESS1; LDS1; LDS1A; LDS2A; MSSE; mutant transforming growth factor beta receptor I; ser; serine/threonine-protein kinase receptor R4; SKR4; TbetaRI; tbetaR-I; tgf beta receptor 1; TGF-beta receptor type I; TGF-beta receptor type-1; TGF-beta type I receptor; TGFBR1; TGFR1; TGFR-1; Transfor; transforming growth factor beta receptor 1; transforming growth factor beta receptor I; transforming growth factor, beta receptor 1; transforming growth factor, beta receptor I; transforming growth factor, beta receptor I (activin A receptor type II-like kinase, 53kDa); transforming growth factor-beta receptor type I | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21812, 29591, 7046 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.92 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P36897, P80204, Q64729 | |
| TGFBR1 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TGF beta Receptor I (TGFBR1) (NP_0046031). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction