missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGFBR3 Monoclonal Antibody (OTI6F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809349
This item is not returnable.
View return policy
Description
This locus encodes the transforming growth factor (TGF)-beta type III receptor. The encoded receptor is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
| TGFBR3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q03167 | |
| TGFBR3 | |
| Human recombit protein fragment corresponding to amino acids 531-777 of human TGFBR3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6F8 | |
| Unconjugated | |
| TGFBR3 | |
| BGCAN, betaglycan | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7049 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction