Learn More
Invitrogen™ TGFBR3 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA5142353
Product Code. 17837662
  • € 465.00 / 100µL
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This locus encodes the transforming growth factor (TGF)-beta type III receptor. The encoded receptor is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TGFBR3
Polyclonal
Unconjugated
TGFBR3
1110036H20Rik; AU015626; AW215636; betaglycan; betaglycan proteoglycan; BGCAN; OTTHUMP00000011787; TBRIII; tgf beta receptor 3; TGF-beta receptor type 3; TGF-beta receptor type III; Tgfbr3; TGFR-3; transforming growth factor beta receptor 3; transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3; transforming growth factor, beta receptor 3; transforming growth factor, beta receptor III
Goat
Ammonium sulfate precipitation
RUO
7049
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q03167
TGFBR3
Peptide with sequence C-TKSIRDDIPSTQGN (aa 313-326).