missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGIF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566463
This item is not returnable.
View return policy
Description
Immunogen sequence: SVLARPSVICH TTVTALKDVP FSLCQSVGVG QNTDIQQIAA KNFTDTSLMY PEDTCKSGPS TNTQ Highest antigen sequence identity to the following orthologs - mouse 82%, rat 77%.
TGIF is a member of the three-amino acid loop extension (TALE) superclass of atypical homeodomains. TALE homeobox proteins are highly conserved transcription regulators. This particular homeodomain binds to a previously characterized retinoid X receptor responsive element from the cellular retinol-binding protein II promoter. In addition to its role in inhibiting 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element, the protein is an active transcriptional co-repressor of SMAD2 and may participate in the transmission of nuclear signals during development and in the adult. Mutations in this gene are associated with holoprosencephaly type 4, which is a structural anomaly of the brain.
Specifications
| TGIF | |
| Polyclonal | |
| Unconjugated | |
| TGIF1 | |
| 5'-TG-3'-interacting factor 1; AA959811; AI462167; Homeobox protein TGIF1; HPE4; TALE family homeobox; TALE homeobox TG-interacting factor; Tfig; TG interacting factor 1; TGFB induced factor homeobox 1; TGFB-induced factor homeobox 1; TGIF; Tgif1; transforming growth factor-beta-induced factor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7050 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15583 | |
| TGIF1 | |
| Recombinant Human TGIF. Recombinant protein control fragment (Product #RP-105224). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction