Learn More
TGIF1 Monoclonal Antibody (OTI4D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802283S
Encompass_Preferred
 View more versions of this product
Product Code. 17570813
  • € 242.00 / 30µL
Description
TGIF1 Monoclonal antibody specifically detects TGIF1 in Human samples. It is validated for Western Blot
Specifications
Specifications
TGIF1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q15583
Mouse
Affinity chromatography
RUO
7050
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI4D10
Unconjugated
TGIF1
5'-TG-3'-interacting factor 1; AA959811; AI462167; Homeobox protein TGIF1; HPE4; TALE family homeobox; TALE homeobox TG-interacting factor; Tfig; TG interacting factor 1; TGFB induced factor homeobox 1; TGFB-induced factor homeobox 1; TGIF; Tgif1; transforming growth factor-beta-induced factor
Full length human recombinant protein of human TGIF1 produced in E.coli.
30 μL