Learn More
TGM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12912-3-AP-20UL
 View more versions of this product
Product Code. 16870864
  • € 145.00 / 20µL
Description
This antibody recognizes the endogenous 90 kDa TGM1 proteins.
The protein encoded by this gene is a membrane protein that catalyzes the addition of an alkyl group from an akylamine to a glutamine residue of a protein, forming an alkylglutamine in the protein. This protein alkylation leads to crosslinking of proteins and catenation of polyamines to proteins. This gene contains either one or two copies of a 22 nt repeat unit in its 3′ UTR. Mutations in this gene have been associated with autosomal recessive lamellar ichthyosis (LI) and nonbullous congenital ichthyosiform erythroderma (NCIE).
Specifications
Specifications
TGM1
Polyclonal
Unconjugated
TGM1
Epidermal TGase, ICR2, KTG, LI, LI1, TG(K), TGASE, TGase 1, TGase K, TGK, TGM1, Transglutaminase 1, Transglutaminase K
Rabbit
Antigen Affinity Chromatography
RUO
21816, 60335, 7051
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P22735, P23606, Q9JLF6
Tgm1
TGM1 Fusion Protein Ag3905
20 μL
Primary
Rat, Human, Mouse