Learn More
Invitrogen™ TGM2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA587544
Product Code. 16586011
  • € 520.00 / 100µL
Description
Positive Samples: SW480, A-549, DU145, PC-3 Immunogen sequence: TNYNSAHDQN SNLLIEYFRN EFGEIQGDKS EMIWNFHCWV ESWMTRPDLQ PGYEGWQALD PTPQEKSEGT YCCGPVPVRA IKEGDLSTKY DAPFVFAEVN ADVVDWIQQD DGSVHKSINR SLIVGLKIST KSVGRDERED ITHTYKYPEG SSEEREAFTR ANHLNKLAEK EETGMAMRIR VGQSMNMGSD FDVFAHITNN TAEEYVCRLL LCARTVSYNG ILGPECGTKY LLNLNLEPFS GKALCSWSIC.
Transglutaminase (TGase) catalyses the crosslink of proteins by forming e-(g-glutamyl) lysine isopeptide bonds and requires the binding of Ca2+ for its activity. In mammals, eight distinct TGase isoenzymes have been identified. Tissue transglutaminase (tTGase), also known as TGase 2, has four distinct domains: N-terminal b-sandwich, catalytic core and two C-terminal b-barrel domains. tTGase may have a role in cell death, cell proliferation, cell differentiation, and receptor-mediated endocytosis. In the Alzheimer's disease brain, the elevated tTGase activity is manifested by polymerization of a number of proteins, including Ab peptide, b-amyloid precursor protein and the tau protein, with formation of neurofibrillary tangles.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TGM2
Polyclonal
Unconjugated
TGM2
C polypeptide; C polypeptide, protein-glutamine-gamma-glutamyltransferase; G alpha h; G[a]h; G-ALPHA-h; GNAH; Protein-glutamine gamma-glutamyltransferase 2; protein-glutamine-gamma-glutamyltransferase; RP5-1054A22.2; TG C; TG(C); TG2; TGase C; TGase H; TGase2; TGase-2; TGase-H; TgaseII; TGC; TGM2; Tissue transglutaminase; Tissue type transglutaminase; tissue-type transglutaminase; transglutaminase; transglutaminase 2; transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase); Transglutaminase 2 C polypeptide; transglutaminase 2, C polypeptide; transglutaminase C; Transglutaminase H; transglutaminase-2; tTG; tTGas
Rabbit
Affinity chromatography
RUO
21817, 56083, 7052
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.73 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
P21980, P21981
TGM2
Recombinant fusion protein containing a sequence corresponding to amino acids 299-548 of human Transglutaminase 2 (TGM2) (NP_9451891).