missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGN46 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49643PCP
This item is not returnable.
View return policy
Description
TGN46 Polyclonal antibody specifically detects TGN46 in Human, Mouse, Bovine, Monkey samples. It is validated for Flow Cytometry, Flow (Intracellular), KnockDown
Specifications
| TGN46 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Bovine, Monkey | |
| Purified |
| Flow Cytometry, KnockDown | |
| PerCP | |
| TGN 46, TGN 48, TGN 51, TGN38, TGN-38, TGN38 homolog, TGN46, TGN-46, TGN48, TGN51, TGOLN 2, TGOLN2, trans golgi network 38, trans golgi network 46, Trans Golgi network integral membrane protein 2, Trans Golgi network integral membrane protein 2 [Precursor], Trans Golgi network protein (46 48 51kD isoforms), Trans golgi network protein 2, Trans Golgi network protein TGN51, Trans-Golgi network integral membrane protein 2, Trans-Golgi network protein 2, Trans-Golgi network protein TGN51, TTGN 2, TTGN2 | |
| A genomic peptide made to an internal region of the human TGN46 protein (within residues 200-350). [Swiss-Prot: O43493] | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Golgi Apparatus Markers | |
| 10618 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction