missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGR5/GPBAR1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-23669R
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TGR5/GPBAR1 Polyclonal specifically detects TGR5/GPBAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Especificaciones
| TGR5/GPBAR1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated | |
| BG37hBG37, G protein-coupled bile acid receptor 1, GPCR19, GPR131, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, M-BARhGPCR19, membrane bile acid receptor, Membrane-type receptor for bile acids, MGC40597, TGR5GPCR | |
| Synthetic peptide from C-terminus of the human TGR5/GPBAR1 protein [UniProt Q8TDU6] | |
| 0.1 mL | |
| GPCR | |
| 151306 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido