missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGR5 Polyclonal Antibody

Description
Sequence homology: Bovine (86%).
This gene encodes a member of the G protein-coupled receptor (GPCR) superfamily. This enzyme functions as a cell surface receptor for bile acids. Treatment of cells expressing this GPCR with bile acids induces the production of intracellular cAMP, activation of a MAP kinase signaling pathway, and internalization of the receptor. The receptor is implicated in the suppression of macrophage functions and regulation of energy homeostasis by bile acids. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
Specifications
| Antigen | TGR5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | GPBAR1 |
| Gene Accession No. | Q80SS6, Q80T02, Q8TDU6 |
| Gene Alias | BG37; C16DUPq36.3; DUP7q36.3; G protein-coupled bile acid receptor 1; G protein-coupled receptor; G protein-coupled receptor TGR5; Gpbar1; GPCR; GPCR HBG37; GPCR19; GPR131; G-protein coupled bile acid receptor 1; G-protein coupled bile acid receptor BG37; G-protein coupled receptor GPCR19; hBG37; Helodermin-preferring VIP receptor; hGPCR19; M-BAR; membrane bile acid receptor; membrane-type receptor for bile acids; PACAP type III receptor; PACAP-R3; PACAP-R-3; pituitary adenylate cyclase-activating polypeptide type III receptor; Tgr5; vasoactive intestinal peptide receptor 2; vasoactive intestinal polypeptide receptor 2; vasopressive intestinal peptide receptor 2; VIP and PACAP receptor 2; VIP receptor subtype 2; Vip2; VIP2R; VIPR2; VIP-R-2; VPAC2; VPAC2 receptor; VPAC2R; VPCAP2R |
| Gene Symbols | GPBAR1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?