missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ TH 257
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
TH 257 is a potent and selective allosteric LIMK 1/2 inhibitor (IC50 values are 84 and 39 nM, respectively). Inhibits cofilin phosphorylation in vitro. Exhibits no significant activity against the wider kinome.
Specifications
Specifications
| Chemical Name or Material | N-Butyl-4-[(phenylamino)sulfonyl]-N-(phenylmethyl)benzamide |
| CAS | 2244678-29-1 |
| Quantity | 25 mg |
| Molecular Formula | C24H26N2O3S |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction