missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TH Monoclonal Antibody (OTI3H3), TrueMAB™, OriGene
Description
TH Monoclonal antibody specifically detects TH in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
| Antigen | TH |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI3H3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | TH |
| Gene Accession No. | P07101 |
| Gene Alias | dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?