missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Th-POK Antibody (PCRP-ZBTB7B-1B6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08311H
This item is not returnable.
View return policy
Description
Th-POK Monoclonal antibody specifically detects Th-POK in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| Th-POK | |
| Monoclonal | |
| HRP | |
| hcKroxc-Krox, Krueppel-related zinc finger protein cKrox, kruppel-related zinc finger protein hcKrox, T-helper-inducing POZ/Krueppel-like factor, THPOK, ZBTB15zinc finger protein 67 homolog (mouse), zfp-67, ZFP67, zinc finger and BTB domain containing 15, zinc finger and BTB domain containing 7B, Zinc finger and BTB domain-containing protein 15, zinc finger and BTB domain-containing protein 7B, Zinc finger protein 67 homolog, Zinc finger protein 857B, Zinc finger protein Th-POK, ZNF857BDKFZp686G01254 | |
| Recombinant full-length human Th-POK protein (Uniprot: O15156) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-ZBTB7B-1B6 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 51043 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction