Learn More
Tocris Bioscience™ Thalidomide 4'-oxyacetamide-PEG1-amine
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mg
missing translation for 'unitSize'
25mg
Descrizione
Thalidomide 4'-oxyacetamide-PEG1-amine is a cereblon ligand for PROTAC® research and development; incorporates a cereblon ligand plus a PEG linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Thalidomide - linker 13
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifica
Specifica
| Chemical Name or Material | N-[2-(2-Aminoethoxy)ethyl]-2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]oxy]acetamide |
| CAS | 2204226-02-6 |
| Synonym | Thalidomide - linker 13 |
| Quantity | 25 mg |
| Molecular Formula | C19H22N4O7.HCl |
| Formula Weight | 454.86 |
| Product Type | Ligand |
| Purity | 95% |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto