missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thalidomide-O-amido-C6-NH2 TFA, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Thalidomide-O-amido-C6-NH2 TFA (Cereblon Ligand-Linker Conjugates 11 TFA), a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker, can be used in the synthesis of PROTACs.
Specifications
Specifications
| Chemical Name or Material | Thalidomide-O-amido-C6-NH2 TFA |
| CAS | 1950635-14-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H27F3N4O8 |
| Quantity | 100 mg |
| Synonym | Cereblon Ligand-Linker Conjugates 11 TFAE3 Ligase Ligand-Linker Conjugates 25 TFA |
| Solubility Information | DMSO : ≥ 105.5 mg/mL (193.76 mM) |
| SMILES | O=C(NCCCCCCN)COC1=CC=CC(C(N2C(CC3)C(NC3=O)=O)=O)=C1C2=O.O=C(O)C(F)(F)F |
| Molecular Weight (g/mol) | 544.48 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction