missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thalidomide-O-COOH, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Thalidomide-O-COOH (Cereblon ligand 3) is the Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Thalidomide-O-COOH (Cereblon ligand 3) can be connected to the ligand for protein by a linker to form PROTACs.
Specifications
Specifications
| Chemical Name or Material | Thalidomide-O-COOH |
| CAS | 1061605-21-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C15H12N2O7 |
| Quantity | 500 mg |
| Synonym | Cereblon ligand 3 E3 ligase Ligand 3 |
| Solubility Information | DMSO : ≥ 42 mg/mL (126.41 mM) |
| SMILES | O=C(N(C(CCC1=O)C(N1)=O)C2=O)C(C2=CC=C3)=C3OCC(O)=O |
| Molecular Weight (g/mol) | 332.26 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction