missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12584-1-AP-20UL
This item is not returnable.
View return policy
Description
12584-1-AP is a rabbit polyclonal antibody raised against full-length of human THAP1.
DNA-binding transcription regulator that regulates endothelial cell proliferation and G1/S cell-cycle progression. Specifically binds the 5′-[AT]NTNN[GT]GGCA[AGT]-3′ core DNA sequence and acts by modulating expression of pRB-E2F cell-cycle target genes, including RRM1. Component of a THAP1/THAP3-HCFC1-OGT complex that is required for the regulation of the transcriptional activity of RRM1. May also have pro-apoptopic activity by potentiating both serum-withdrawal and TNF-induced apoptosis.Specifications
| THAP1 | |
| Polyclonal | |
| Unconjugated | |
| THAP1 | |
| THAP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 306547, 55145, 73754 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5U208, Q8CHW1, Q9NVV9 | |
| THAP1 | |
| THAP1 Fusion Protein Ag3285 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction