missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ THAP11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16380545
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16380545 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16380545 Supplier Invitrogen™ Supplier No. PA583593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QQQQSSPSAS TAQTAQLQPN LVSASAAVLL TLQATVDSSQ APGSVQPAPI TPTGEDVKPI DLTVQVEFAA AE Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 88%.

This gene encodes two proteins, the SART1protein expressed in the nucleus of the majority of proliferating cells, and the SART1protein expressed in the cytosol of epithelial cancers. The SART1protein is translated by the mechanism of -1 frameshifting during posttranscriptional regulation; its full-length sequence is not published yet. The two encoded proteins are thought to be involved in the regulation of proliferation. Both proteins have tumor-rejection antigens. The SART1protein possesses tumor epitopes capable of inducing HLA-A2402-restricted cytotoxic T lymphocytes in cancer patients. This SART1antigen may be useful in specific immunotherapy for cancer patients and may serve as a paradigmatic tool for the diagnosis and treatment of patients with atopy. The SART1protein is found to be essential for the recruitment of the tri-snRNP to the pre-spliceosome in the spliceosome assembly pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THAP11
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THAP11
Gene Accession No. Q96EK4
Gene Alias 2810036E22Rik; AB041579; CTG-B43a; CTG-B45d; HRIHFB2206; MNCb-2032; Ronin; THAP domain containing 11; THAP domain-containing protein 11; Thap11
Gene Symbols THAP11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human THAP11. Recombinant protein control fragment (Product #RP-98751).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57215
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.