missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ THAP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16656834
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16656834 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16656834 Supplier Invitrogen™ Supplier No. PA5100351

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total THAP2.

THAP2, also named as THAP domain-containing protein 2, is a 228 amino acid protein, which contains one THAP-type zinc finger. Members of the THAP (thanatos-associated protein) family of proteins contain a well conserved DNA-binding domain known as the THAP-type zinc finger motif. Proteins containing the THAP-type zinc finger motif are commonly involved in transcriptional regulation, cell-cycle control, apoptosis and chromatin modification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THAP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene THAP2
Gene Accession No. Q9D305, Q9H0W7
Gene Alias 2900040O07Rik; 9030625G08Rik; AI450385; AI649097; THAP domain containing 2; THAP domain containing, apoptosis associated protein 2; THAP domain-containing protein 2; Thap2
Gene Symbols THAP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human THAP2(Accession Q9H0W7), corresponding to amino acid residues K87-R137.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66816, 688019, 83591
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.