missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20853-1-AP-150UL
This item is not returnable.
View return policy
Description
THAP2, also named as THAP domain-containing protein 2, is a 228 amino acid protein, which contains one THAP-type zinc finger. Members of the THAP (thanatos-associated protein) family of proteins contain a well conserved DNA-binding domain known as the THAP-type zinc finger motif. Proteins containing the THAP-type zinc finger motif are commonly involved in transcriptional regulation, cell-cycle control, apoptosis and chromatin modification.Specifications
| THAP2 | |
| Polyclonal | |
| Unconjugated | |
| THAP2 | |
| THAP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66816, 688019, 83591 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D305, Q9H0W7 | |
| Thap2 | |
| THAP2 Fusion Protein Ag14775 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction