missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THAP6 Antibody (OTI1F6), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74497AF532
This item is not returnable.
View return policy
Description
THAP6 Monoclonal specifically detects THAP6 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| THAP6 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| THAP6 | |
| Full length recombinant protein of human THAP6 (NP_653322) produced in E.coli. | |
| 0.1 mL | |
| 152815.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1F6 | |
| Western Blot, Immunohistochemistry | |
| MGC30052, THAP domain containing 6, THAP domain-containing protein 6 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction