missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THAP6 Monoclonal Antibody (OTI1E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800245S
This item is not returnable.
View return policy
Description
THAP6 Monoclonal antibody specifically detects THAP6 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| THAP6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92552 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23107 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI1E10 | |
| Unconjugated | |
| Mrps27 | |
| THAP domain containing 6; THAP domain-containing protein 6; THAP6 | |
| Human recombinant protein fragment corresponding to amino acids 151-414 of human MRPS27 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction