Learn More
THAP6 Monoclonal Antibody (OTI3A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800248
Encompass_Preferred
 View more versions of this product
Product Code. 17527922
  • € 540.87 / 100µL
Description
THAP6 (THAP domain-containing protein 6) is a 222 amino acid protein that contains one THAP-type zinc finger and exists as two alternatively spliced isoforms. The gene that encodes THAP6 consists of approximately 36,528 bases and maps to human chromosome 4q21.1. Chromosome 4 represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington′s disease, is encoded by a gene that maps to chromosome 4. FGFR-3 is also encoded by a gene located on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also tied to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.
Specifications
Specifications
THAP6
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8TBB0
THAP6
Full length human recombit protein of human THAP6 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI3A8
Unconjugated
THAP6
THAP domain containing 6
Mouse
Affinity Chromatography
RUO
152815
-20° C, Avoid Freeze/Thaw Cycles