Learn More
THAP6 Monoclonal Antibody (OTI3A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800248S
Encompass_Preferred
 View more versions of this product
Product Code. 17547922
  • € 242.00 / 30µL
Description
THAP6 Monoclonal antibody specifically detects THAP6 in Human samples. It is validated for Western Blot
Specifications
Specifications
THAP6
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8TBB0
Mouse
Affinity chromatography
RUO
152815
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI3A8
Unconjugated
THAP6
THAP domain containing 6; THAP domain-containing protein 6; THAP6
Full length human recombinant protein of human THAP6 produced in E.coli.
30 μL