missing translation for 'onlineSavingsMsg'
Learn More

THAP6 Monoclonal Antibody (OTI3B4), TrueMAB™, OriGene

Product Code. p-200016376
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17517922 30 μL 30µL
17597912 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17517922 Supplier Origene Technologies Supplier No. TA800247S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

THAP6 Monoclonal antibody specifically detects THAP6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen THAP6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene THAP6
Gene Accession No. Q8TBB0
Gene Alias THAP domain containing 6; THAP domain-containing protein 6; THAP6
Host Species Mouse
Immunogen Full length human recombinant protein of human THAP6 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152815
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.