Learn More
THAP6 Mouse anti-Human, Clone: OTI4D4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800281
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
THAP6 (THAP domain-containing protein 6) is a 222 amino acid protein that contains one THAP-type zinc finger and exists as two alternatively spliced isoforms. The gene that encodes THAP6 consists of approximately 36,528 bases and maps to human chromosome 4q21.1. Chromosome 4 represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington′s disease, is encoded by a gene that maps to chromosome 4. FGFR-3 is also encoded by a gene located on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also tied to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.Specifications
| THAP6 | |
| Monoclonal | |
| Unconjugated | |
| THAP6 | |
| THAP domain containing 6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 152815 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4D4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8TBB0 | |
| THAP6 | |
| Full length human recombit protein of human THAP6 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.