missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THAP8 Monoclonal Antibody (OTI5A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811429
This item is not returnable.
View return policy
Description
THAP8 is a protein coding gene.Specifications
| THAP8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NA92 | |
| THAP8 | |
| Human recombit protein fragment corresponding to amino acids 86-274 of human THAP8 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5A1 | |
| Unconjugated | |
| THAP8 | |
| THAP domain containing 8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 199745 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction