Learn More
Abeba™ The Original 5081 Shoes
Product Code. 15671808
Click to view available options
Size:
35
36
38
39
40
41
42
43
Unit Size:
1 pair
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15671808
Brand: Abeba™ undefined
Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Animal fats, oil and fuel resistant. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • White/mint
  • Slip resistance outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather top layer
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color White
Material Leather
Size 42
Sole Features Slip-resistant
Style 5081
Resistant To Animal fats, oil and fuel