Learn More
Abeba™ The Original 7081 Shoes
Product Code. 15672508
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
Unit Size:
1 pair
Product Code. 15672508
Brand: Abeba™ 7081/36
Only null left
Add to basket
Add to basket
Animal fats, oil and fuel resistant. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB FO SRC
  • Smooth leather
  • Light blue/White
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB FO SRC
Color Light Blue, White
Material Leather
Size 36
Sole Features Slip-resistant
Style 7081
Resistant To Animal fats, oil and fuel