Learn More
Invitrogen™ THEGL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139879
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DGQVSTEIST CSEVFQKPIV LRILDTHREL EESEDPEKHE NPEEPEEVRE QDQRDESEEC DEPHESYEPH APYAPHKPRD SYAPYELHG Highest antigen sequence indentity to the following orthologs: Mouse - 28%, Rat - 28%.
Specifications
| THEGL | |
| Polyclonal | |
| Unconjugated | |
| THEGL | |
| testicular haploid expressed gene protein-like; theg spermatid protein-like; theg spermatid-like protein; THEGL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100506564 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0DJG4 | |
| THEGL | |
| Recombinant protein corresponding to Human THEGL. Recombinant protein control fragment (Product #RP-109108). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.