missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THEM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97054-100ul
This item is not returnable.
View return policy
Description
THEM2 Polyclonal antibody specifically detects THEM2 in Human samples. It is validated for ELISA
Specifications
| THEM2 | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| acyl-CoA thioesterase 13PNAS-27, EC 3.1.2.-, HT012, hypothalamus protein HT012, MGC4961, THEM2, Thioesterase superfamily member 2acyl-coenzyme A thioesterase 13 | |
| Produced in rabbits immunized with purified, recombinant Human THEM2 (Accession#: NP_060943.1; Met1-Asn140) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 55856 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction