missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Themis Antibody (701010), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6929N
This item is not returnable.
View return policy
Description
Themis Monoclonal antibody specifically detects Themis in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Themis | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 387357 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 701010 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA | |
| E. coli-derived recombinant human Themis, Ala2-Pro282, Accession # Q8N1K5 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction