missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Themis Antibody (701010), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6929E
This item is not returnable.
View return policy
Description
Themis Monoclonal antibody specifically detects Themis in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Themis | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 387357 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 701010 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA | |
| E. coli-derived recombinant human Themis, Ala2-Pro282, Accession # Q8N1K5 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction