missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Themis Antibody (719945), DyLight 755, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB6816Z
This item is not returnable.
View return policy
Description
Themis Monoclonal antibody specifically detects Themis in Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Themis | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 387357 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 719945 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA | |
| E. coli-derived recombinant mouse Themis, Lys122-Lys237, Accession # Q8BGW0 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur