missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Themis Antibody (SR2184), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Themis |
|---|---|
| Clone | SR2184 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650605
|
Novus Biologicals
NBP3-22411-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618554
|
Novus Biologicals
NBP3-22411-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
Themis Monoclonal antibody specifically detects Themis in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationTekniske data
| Themis | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 387357 | |
| IgG | |
| Affinity purified |
| SR2184 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA | |
| A synthesized peptide derived from Themis (Uniprot #: Q8N1K5) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel