Learn More
Theophylline-13C2d6, TRC
Product Code. 30449533
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449533 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30449533 Supplier Toronto Research Chemicals Supplier No. TRCT3438541MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Theophylline-13C2d6
CAS 1782458-84-7
Molecular Formula C513C2H2D6N4O2
Quantity 1 mg
Synonym 3,9-Dihydro-1,3-dimethyl-1H-purine-2,6-dione-13C2d6, 3,7-Dihydro-1,3-dimethyl- 1H-purine-2,6-dione-13C2d6, 1,3-Dimethylxanthine-13C2d6, Accurbron-13C2d6, Aerobin-13C2d6, Armophylline-13C2d6, Asmax-13C2d6, Bronkodyl-13C2d6, Theophyl-13C2d6, Theoplus-13C2d6, Theostat-13C2d6, Unifyl-13C2d6, Unilong-13C2d6, Xanthium-13C2d6
InChI Formula InChI=1S/C7H8N4O2/c1-10-5-4(8-3-9-5)6(12)11(2)7(10)13/h3H,1-2H3,(H,8,9)/i1+1D3,2+1D3
SMILES O=C1C2=C(N=CN2)N([13C]([2H])([2H])[2H])C(N1[13C]([2H])([2H])[2H])=O
Molecular Weight (g/mol) 188.19
Formula Weight 188.1091