Learn More
Heidolph™ Thermal Fluid - Kryo 30 Liquid
Shop All Heidolph Instruments Products
Click to view available options
For Use With (Equipment):
Hei-CHILL Chillers
Unit Size:
20 L
Description
- Non-flammable thermal fluid for heating and low-temperature range
- 20 L thermal fluid for Hei-CHILL Chillers
Specifications
Specifications
| Capacity (Metric) | 20 L |
| For Use With (Equipment) | Hei-CHILL Chillers |
Safety and Handling
Product Identifier
- Kryo 30
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands and face thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethylene glycol