Learn More
Heidolph™ Thermal Fluid - Kryo 30 Liquid
Product Code. 17195354 Shop All Heidolph Instruments Products
Click to view available options
For Use With (Equipment):
Hei-CHILL Chillers
Unit Size:
20 L
Product Code. 17195354
Brand: Heidolph™ 5690061000
Non-flammable thermal fluid for heating and low-temperature range.
  • Non-flammable thermal fluid for heating and low-temperature range
  • 20 L thermal fluid for Hei-CHILL Chillers
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 20 L
For Use With (Equipment) Hei-CHILL Chillers
compliance-icons compliance-icons
Product Identifier
  • Kryo 30
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands and face thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethylene glycol