Learn More
Kern Sohn Thermal Printer
Click to view available options
:
(L x W x H) 140 x 165 x 50 mm
Description
- For printing weighing results, recipes, quantities etc. or for GLP record keeping of balance adjustment with date and time print
- FLP/GMP compliant print form available as standard. Language versions: DE, GB, FR
Specifications
Specifications
| Connection Type | Data Interface RS-232 |
| Electrical Requirements | Mains Adapter External, 100/240 V |
| Dimensions (L x W x H) | 140 x 165 x 50 mm |