Learn More
Brady™ Thermal Transfer Printable Labels
Product Code. p-7144728
Click to view available options
Width (Metric):
63 mm
83 mm
110 mm
Color:
Blue
Green
Red
White
Yellow
Unit Size:
1 roll
Product Code. 11508684
Brand: Brady™ 217509
Only null left
Add to basket
Add to basket
Brady™ Thermal Transfer Printable Labels are comprised of colored vinyl film available in different widths with a permanent acrylic adhesive.
  • Colored, printable vinyl film for thermal transfer printers
  • Permanent acrylic adhesive
  • For general and industrial identification
  • Compatible with Brady printer models BBP12; BBP11; PR200Plus; PR300Plus; PR600Plus; BP1244; BP1344; BP4000; BP4320
  • Supplied as 40m rolls in three widths
  • Service temperatures -70°C to 40°C
Specifications
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Gloss
Format Continuous
Material Pressure-sensitive Vinyl
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Barcode labelling, Component and equipment labelling, Data and telecommunications labelling, Electrical labelling, General identification, Inventory and inspection labelling, Laboratory labelling
For Use With (Equipment) BBP11, BBP12, PR Plus, THT 25 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -70°C
No. per Pack 1 Roll/Pack
Temperature Range (English) -94°F to +176°F
Temperature Range (Metric) -70°C to +80°C
Thickness (English) 0.0042 in.
Thickness (Metric) 0.107 mm
Dimensions (LxW) 40 x 110 mm
Length (English) 131 ft.
Length (Metric) 40 m
Width (English) 4.331 in.
Width (Metric) 110 mm
Alternate Part Number BPTC-110-439-WT