Learn More
Amarell™ Thermo-Hygrometer With Outdoor Probe
Product Code. 10339620
Click to view available options
Unit Size:
Each
Product Code. 10339620
Brand: Amarell™ E915000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use 3m cable for outdoor readings. The Thermo-Hygrometer provides readings between -50 to 70°C and 20 to 90% rF.
Universal thermo-hygrometer with outdoor probe.
  • Accessory for wall mounting (screws or adhesive tape).
  • Indoor/Outdoor
  • Min/Max Function
  • °C ↔ °F
  • With Clock
  • Fast responding semi-conductor probe
  • Probe includes 3m cable for outdoor measurements
Specifications
Type Thermohygrometer
Humidity Range 20 to 90%
Resolution 1% rF (rh)
Accuracy ±5% rF (rh)
Temperature Range (Metric) -50°C to +70°C
Temperature Accuracy ±1°C
Temperature Resolution 0.1°C
Dimensions (L x W x H) 110 x 60 x 20 mm
Length (Metric) Cable 3 m
Voltage 1.5 V
Includes 3m cable and probe