Learn More
Integra Biosciences™ Thermo Plastic Elastomer Micronic capcluster-96
Product Code. 10391860
Encompass
Change view
Click to view available options
Material:
TPE
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10391860 TPE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10391860 Supplier Integra Biosciences™ Supplier No. MP53041
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Specifications
Color Assorted
Material TPE
Autoclavable Autoclavable
Format SBS 8X12 Microplate Standard
Product Type Capcluster
Pierceable Yes