Learn More
Thermo Scientific™ Barnstead™ Easypure™ II Compact Ultrapure Water System Filters and Cartridges
Product Code. 10669532 Shop All Thermo Scientific Products
Change view
Click to view available options
Resin Type:
Activated Carbon and Mixed Bed Ion Exchange Resin
Ultrapure Mixed Bed
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Resin Type unitSize
10669532 Activated Carbon and Mixed Bed Ion Exchange Resin Each
10032283 Ultrapure Mixed Bed Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10669532 Supplier Thermo Scientific™ Supplier No. D50231
  • Easypure™ II Systems require pretreated feedwater
  • From the chart, select appropriate cartridges based on application and feedwater type
Order Info
Cartridges and filters must be ordered separately
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Pretreated
Resin Type Activated Carbon and Mixed Bed Ion Exchange Resin
For Use With (Application) Organic/ion exchange cartridge
For Use With (Equipment) Easypure II Compact Ultrapure Water System, Organic/Ion Exchange Cartridge
Type Cartridge and Filter