Learn More
Thermo Scientific™ Barnstead™ Easypure™ II Compact Ultrapure Water System Filters and Cartridges
Shop All Thermo Scientific ProductsDescription
- Easypure™ II Systems require pretreated feedwater
- From the chart, select appropriate cartridges based on application and feedwater type
Order Info
Cartridges and filters must be ordered separately
Specifications
Specifications
| Feedwater Source | Pretreated |
| Resin Type | Activated Carbon and Mixed Bed Ion Exchange Resin |
| For Use With (Application) | Organic/ion exchange cartridge |
| For Use With (Equipment) | Easypure II Compact Ultrapure Water System, Organic/Ion Exchange Cartridge |
| Type | Cartridge and Filter |